Would you like to log in again?
MDA-oxLDL ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
BI-20022
102993-974EA 817.31 USD InStock
102993-974
MDA-oxLDL ELISA, Eagle Biosciences
Assays ELISAs
MDA Oxidized LDL EIA is intended to detect human MDA Oxidized LDL concentrations in serum or plasma.
  • Dynamic Range: 0 - 10 µg/ml
  • Sensitivity: 0.05 µg/ml
  • Incubation: 3.5 hours
The Eagle Biosciences MDA Oxidized LDL ELISA Assay kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of human MDA Oxidized LDL in serum or plasma. The MDA Oxidized LDL ELISA Assay kit is manufactured by Biomedica.  The MDA Oxidized LDL ELISA Assay kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty