Would you like to log in again?
oLAB ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
BI-20032
102993-976EA 1000.38 USD InStock
102993-976
oLAB ELISA, Eagle Biosciences
Assays ELISAs
OLAB IgG ELISA detects human IgG autoantibodies against oxidized low density lipoprotein in serum.
  • Dynamic Range: 37 - 1,200 mU/ml
  • Sensitivity: 37 mU/ml
  • Incubation: 2.5 hours
Oxidized low density lipoprotein (oLDL) is believed to play a critical role in the development and progression of atherosclerosis. Accumulation of oLDL in macrophages and smooth muscle cells causes foam cell formation, an initial step in the disease. Autoantibodies against oxidatively modified LDL can be used as a parameter that consistently mirrors the occurrence of oxidation processes taking place in vivo. In fact, elevated levels of autoantibodies against oLDL have been detected in the blood stream of patients with coronary artery disease. Moreover, recent studies indicate a correlation between autoantibodies against oLDL and the progression of carotid atherosclerosis. Increased serum concentrations of oLAB have also been described in various diseases such as pre-eclampsia and systemic lupus erythematosus. Decreased oLAB titers were observed during septicemia and myocardial infarction.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty