Would you like to log in again?
Big Endothelin ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
BI-20082H
102993-980EA 941.54 USD InStock
102993-980
Big Endothelin ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Human Big Endothelin-1 ELISA detects Big Endothelin-1 in serum & plasma.
  • Dynamic Range: 0.3 - 12 ng/ml
  • Sensitivity: 0.01 ng/ml
  • Incubation: 1.5 hours
Big Endothelin-1 (BigET) is a peptide of 38 amino acids and is the precursor of Endothelin-1 (ET), represented by amino acids 1-21. ET is a potent vasoconstrictor and is produced by vascular endothelial cells. Accordingly it has a wide tissue distribution. The cleavage of BigET by Endothelin Converting Enzyme (ECE) leads to ET and to a C‑terminal fragment. Both BigET and ET are strong independent predictors of survival in patients with congestive heart failure, and identify a population with a very high risk mortality. The half-life of ET (1-21) in plasma is less than one minute, whereas clearance of BigET is much slower. BigET can therefore be determined more easily.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty