Would you like to log in again?
OPG ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
BI-20403
102993-982EA 1078.85 USD InStock
102993-982
OPG ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human Osteoprotegerin ELISA detects human osteoprotegerin levels in serum or plasma samples.
  • Dynamic Range: 0 to 20 pmol/l
  • Sensitivity: LOD: (0 pmol/l + 3 SD): 0.07 pmol/l; LLOQ: 0.08 pmol/l
  • Incubation: 5.5 hours
Osteoprotegerin (OPG) or Osteoclast inhibitory factor (OCIF) is a glycoprotein of the TNF receptor superfamily 11b (gene name TNFRSF11B). OPG is synthesized as a monomer of 380 amino acids and is assembled as a homodimer within the cell and then secreted mainly as a disulfide-linked homodimer into the extracellular compartment. OPG is produced by many different tissues and cell types including osteoblasts. OPG is a negative regulator of bone resorption by acting as decoy receptor for RANKL, thus neutralizing its function in osteoclastogenesis. This glycoprotein is also involved in the regulation of vascular calcification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty