Would you like to log in again?
FGF23 (C-terminal) ELISA Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
BI-20702
102993-984EA 935 USD InStock
102993-984
FGF23 (C-terminal) ELISA Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
FGF23 (Fibroblast Growth Factor 23) ELISA detects FGF23 in serum or plasma.
  • Dynamic Range: 0.63 - 10 nmol/L
  • Sensitivity: 0.05 nmol/L
  • Incubation: 3.5 hours
FGF23 (fibroblast growth factor 23) is a 32 kDa protein with 251 amino acids that is proteolytically processed between arginine179 and serine180 to generate N-terminal and C-terminal fragments. FGF23 is mainly secreted by osteocytes and controls phosphate and 1,25(OH)2 vitamin D homeostasis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty