Would you like to log in again?
Endostatin ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
BI-20742
102993-986EA 928.46 USD InStock
102993-986
Endostatin ELISA, Eagle Biosciences
Assays ELISAs
Endostatin ELISA detects human Endostatin concentrations in serum and plasma.
  • Dynamic Range: 0.2 - 20 pmol/L
  • Sensitivity: 0.1 pmol/L
  • Incubation: Overnight
Endostatin, a 20-kDa C-terminal proteolytic fragment of collagen XVIII, is an endogenous angiogenesis inhibitor localized in the vascular basement membrane in various organs.  The biological functions of the endostatin-network involve SPARC, thrombospondin-1, glycosaminoglycans, collagens, and integrins.   Endostatin is expressed during the progression of renal fibrosis in tubular cells of injured tissue. In renal micro-vascular disease, observed in late stages of patients with chronic kidney disease, increased endostatin levels are possibly the consequence of enhanced extracellular matrix degradation. Thus endostatin may become an important marker for progressive microvascular renal disease in patients with chronic kidney disease. Endostatin levels in blood are also likely to increase in patients with other microvascular tissue injuries, including atherosclerosis, myocardial- and brain ischemia. In ischemic stroke patients, high endostatin plasma levels predict a worse long-term clinical outcome.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty