Would you like to log in again?
EZ4U Cell Proliferation Assay, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
BI-5000
102993-994EA 385.77 USD InStock
102993-994
EZ4U Cell Proliferation Assay, Eagle Biosciences, Inc.
Assays ELISAs
EZ4U (Nonradioactive Cell Proliferation and Cytoxicity) ELISA from sm Inc
  • Dynamic Range: 15 -960 U/mL
  • Sensitivity: 1 U/mL
  • Incubation: 2 hours
Proliferation assays are widely used in cell biology for the study of growth factors, cytokines, nutrients and for the screening of cytotoxic or chemotherapeutic agents. There are several ways to determine the number of cells either by microscopic inspection, or by the use of an electronic particle counter, indirectly by measuring the incorporation of radioactive precursors, quantitating total protein with chromogenic dyes, or by measuring metabolic activity of cellular enzymes. The most common assay for cell proliferation is the incorporation of 3H‑thymidine into cellular DNA. The 3H-thymidine assay is, however, labour intensive as it requires the removal of excess, unincorporated label by using some method of cell harvesting before measurement. In 1956, the first paper was published on the use of tetrazolium salts as indicators of cell viability. The method was based on the finding that living cells are capable to reduce slightly or uncoloured tetrazolium salts into intensely coloured formazan derivatives. This reduction process requires functional mitochondria, which are inactivated within a few minutes after cell death.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty