Would you like to log in again?
OxyStat ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
BI-5007
102993-996EA 921.92 USD InStock
102993-996
OxyStat ELISA, Eagle Biosciences
Assays ELISAs
OxyStat Colorimetric detects peroxides in plasma, serum and cell culture supernatants.
  • Dynamic Range: 7 - 600 µmol/l
  • Sensitivity: 7 µmol/l
  • Incubation: 15 minutes
Cells and tissues are sensitive to oxidative stress, caused by the formation of free radicals. If not deactivated by antioxidants, organic peroxides and hydroperoxides are the first reaction products between cellular constituents and free radicals or other reactive oxygen derivates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty