Would you like to log in again?
Biotin ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
BIO06-K01
102993-998EA 1124.62 USD InStock
102993-998
Biotin ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Biotin ELISA is intended to detect Biotin in food products by enzyme linked immuno .
  • Dynamic Range: 1.0 - 25 ng/ml
  • Sensitivity: 0.5 ng/mL
  • Incubation: 1.5 hours
Biotin serves as the prosthetic group of enzymes, which catalyze carboxylations in the organism. For this purpose, biotin is bound via its carboxy group to lysin residues of carboxylases, and the transfer of carbon dioxide takes place after its attachment to a nitrogen atom of biotin, forming the so-called active carbon dioxide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty