Would you like to log in again?
Human CD106 / VCAM-1 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C0631-K01
102994-026EA 830.38 USD InStock
102994-026
Human CD106 / VCAM-1 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human sVCAM-1/CD106 ELISA detects soluble sVCAM-1 in various samples.
  • Dynamic Range: 15.625 - 1000 ng/ml
  • Sensitivity: 156 ng/ml (sample dilution 1:10)
  • Incubation: 2.5 hours
The Eagle Biosciences Human sVCAM-1/CD106 ELISA is a solid phase sandwich ELISA for the in-vitro qualitative and quantitative determination of soluble Vascular Cellular Adhesion Molecule-1 (sVCAM-1) in cell culture supernatants, buffered solutions or human serum, plasma, or other body fluids. This assay will recognize both natural and recombinant human sVCAM-1. The Eagle Biosciences Human sVCAM-1/CD106 ELISA Assay kit is for Research Use Only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty