Would you like to log in again?
Human CD14 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C1431-K01
102994-028EA 830.38 USD InStock
102994-028
Human CD14 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human CD14 ELISA determines the in-vitro CD14 in serum, plasma, or cell lysates.
  • Dynamic Range: 6.87 - 220 ng/ml
  • Sensitivity: 6 ng/mL
  • Incubation: 3.75 hours
The Eagle Biosciences Human CD14 ELISA Assay kit is to be used for the in-vitro quantitative determination of CD14 in serum, plasma, or cell lysates. The assay will recognize both natural and recombinant CD14. The Eagle Biosciences Human CD14 ELISA Assay kit is for Research Use Only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty