Would you like to log in again?
Human CD116 / GM-CSF R ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C1631-K01
102994-030EA 830.38 USD InStock
102994-030
Human CD116 / GM-CSF R ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human CD116 ELISA determines CD116 in serum, plasma, or cell culture supernatants.
  • Dynamic Range: 31.25 pg/ml - 1000 pg/ml
  • Sensitivity: 16 pg/ml
  • Incubation: 3.75 hours
The Eagle Biosciences Human soluble GM-CSF receptor α chain (CD116) ELISA is to be used for the in-vitro quantitative determination of CD116 in serum, plasma, or cell culture supernatants. The assay will recognize both natural and recombinant CD116. The Eagle Biosciences Human CD116 ELISA Assay kit is for Research Use Only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty