Would you like to log in again?
Human CD117 / c-KIT ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C1731-K01
102994-032EA 830.38 USD InStock
102994-032
Human CD117 / c-KIT ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human CD117 ELISA detects Stem Cell Factor receptor molecule CD117 in biological fluids.
  • Dynamic Range: 0.312 - 10 ng/ml
  • Sensitivity: 0.095 ng/ml
  • Incubation: 1.75 hours
The Eagle Biosciences Human CD117 ELISA is to be used for the in-vitro quantitative determination of Stem Cell Factor receptor molecule (CD117) in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant CD117. The Eagle Biosciences Human CD117 ELISA Assay kit is for Research Use Only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty