Would you like to log in again?
Human CD124 / IL-4R ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C2431-K01
102994-036EA 830.38 USD InStock
102994-036
Human CD124 / IL-4R ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human IL-4R ELISA detects soluble IL-4 receptor molecule (CD124) in various samples.
  • Dynamic Range: 31.25-1000 pg/ml
  • Sensitivity: 11.5 pg/ml
  • Incubation: 1h45
The Eagle Biosciences Human IL-4R ELISA is to be used for the in-vitro quantitative determination of  soluble IL-4 receptor molecule (CD124)  in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant human IL-4R.  The Eagle Biosciences Human IL-4R ELISA Assay kit is for Research Use Only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty