Would you like to log in again?
Human CD62L / LAM-1 / L-Selectin ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C2L31-K01
102994-044EA 830.38 USD InStock
102994-044
Human CD62L / LAM-1 / L-Selectin ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human CD62L ELISA detects CD62L in human serum, plasma, buffered solutions or cell culture.
  • Dynamic Range: 1.6-60 ng/ml
  • Sensitivity: 1 ng/ml
  • Incubation: 1h45
The Eagle Biosciences Human CD62L ELISA is to be used for the in-vitro quantitative determination CD62L in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant human CD62L. The Eagle Biosciences Human CD62L ELISA Assay kit is for Research Use Only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty