Would you like to log in again?
Human CD62P / P-Selectin ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C2P31-K01
102994-046EA 830.38 USD InStock
102994-046
Human CD62P / P-Selectin ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human CD62P ELISA is used for the quantitative determination of CD62P, in biological samples.
  • Dynamic Range: 4.4-140 ng/ml
  • Sensitivity: 3.6 ng/ml
  • Incubation: 2h45
A soluble form of P-selectin, which might represent a proteolytic fragment or a soluble splice variant lacking the transmembrane domain, is found in serum and plasma (2). High expression of P-selectin or sP-selectin has been implicated in several inflammatory disorders, including adult respiratory distress syndrome, acute lung injury, ischemia-reperfusion injury, Gram-negative septic shock, thrombotic diseases, malaria, systemic sclerosis, connective tissue disease and rheumatoid arthritis. Furthermore, lung injury scores correlate significantly with sP-selectin in plasma of patients with acute lung injury (3) and (4).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty