Would you like to log in again?
Human CD138 / SYNDECAN-1 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C3831-K01
102994-052EA 830.38 USD InStock
102994-052
Human CD138 / SYNDECAN-1 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
sCD138 ELISA is to detect sCD138 in supernatants, buffered solutions or serum and plasma samples.
  • Dynamic Range: 8-256 ng/ml
  • Sensitivity: 2.56 ng/ml
  • Incubation: 1h45
The Eagle Biosciences sCD138 ELISA kit is a solid phase sandwich ELISA for the in-vitro qualitative and quantitative determination of sCD138 in supernatants, buffered solutions or serum and plasma samples. This assay will recognise both natural and recombinant human sCD138.  The Eagle Biosciences Human sCD138 ELISA Assay kit is for Research Use Only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty