Would you like to log in again?
Human CD44 / Pgp-1 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C4431-K01
102994-056EA 830.38 USD InStock
102994-056
Human CD44 / Pgp-1 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human CD44 ELISA detects CD44 in human serum, plasma, buffered solutions or cell culture medium.
  • Dynamic Range: 0.31-10 ng/ml
  • Sensitivity: 11 ng/ml
  • Incubation: 2h10
The Eagle Biosciences CD44 ELISA is to be used for the in-vitro quantitative determination of soluble CD44 (CD44/phagocytic glycoprotein-1/Pgp-1) in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant human CD44. The Eagle Biosciences Human CD44 ELISA Assay kit is for Research Use Only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty