Would you like to log in again?
Human CD50 / ICAM-3 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C5031-K01
102994-060EA 830.38 USD InStock
102994-060
Human CD50 / ICAM-3 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
sICAM-3/CD50 ELISA detects soluble Inter Cellular Adhesion Molecule -3 in biological samples.
  • Dynamic Range: 0.625-20 ng/ml
  • Sensitivity: 0.3 ng/ml
  • Incubation: 1h40
The Eagle Biosciences sICAM-3 / CD50 ELISA is to be used for the in-vitro quantitative determination of soluble Inter Cellular Adhesion Molecule -3 (sICAM-3) in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant human sICAM-3. The Eagle Biosciences Human CD50 / sICAM-3 ELISA Assay kit is for Research Use Only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty