Would you like to log in again?
Human CD54 / ICAM-1 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C5431-K01
102994-064EA 830.38 USD InStock
102994-064
Human CD54 / ICAM-1 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human sICAM-1/CD54 EIA detects soluble Inter Cellular Adhesion Molecule -1 in various sample types.
  • Dynamic Range: 0.25-8 ng/ml
  • Sensitivity: 0.1 ng/ml
  • Incubation: 1h40
The Eagle Biosciences Human sICAM-1/CD54 ELISA is to be used for the in-vitro quantitative determination of soluble Inter Cellular Adhesion Molecule -1 (sICAM-1) in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant human sICAM-1/CD54. The Eagle Biosciences Human sICAM-1/CD54 ELISA Assay kit is for Research Use Only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty