Would you like to log in again?
Human Trail Receptor 1/DR4/CD261 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C6131-K01
102994-066EA 830.38 USD InStock
102994-066
Human Trail Receptor 1/DR4/CD261 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human TRAIL R1/DR4 ELISA detects TRAIL R1/DR4 in biological fluids.
  • Dynamic Range: 31.25-1000 pg/ml
  • Sensitivity: 8 pg/ml
  • Incubation: 2h30
The Eagle Biosciences Human TRAIL R1/DR4 ELISA is a solid phase sandwich ELISA for the in-vitro qualitative and quantitative determination of TRAIL R1/DR4 in cell culture supernatants, buffered solutions or human serum, plasma, or other body fluids. This assay will recognize both natural and recombinant human TRAIL R1/DR4. The Eagle Biosciences Human TRAIL R1/DR4 ELISA Assay kit is for Research Use Only. 
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty