Would you like to log in again?
Human CD178 / FAS-Ligand ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C7831-K01
102994-074EA 830.38 USD InStock
102994-074
Human CD178 / FAS-Ligand ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human soluble CD178/ Fas-L/CD95-L EIA detects the soluble human Fas Ligand in various sample types.
  • Dynamic Range: 62.5-2000 pg/ml
  • Sensitivity: 12 pg/ml
  • Incubation: 3h45
The Eagle Biosciences Human soluble CD178/ Fas-L/CD95-L ELISA is to be used for the in-vitro quantitative determination of soluble human Fas Ligand (Fas-L) in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant human Fas-L. The Eagle Biosciences Human CD178 ELISA Assay kit is for Research Use Only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty