Would you like to log in again?
Human CD95 / FAS ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C9531-K01
102994-078EA 830.38 USD InStock
102994-078
Human CD95 / FAS ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human soluble CD95 ELISA detects soluble CD95 in biological samples.
  • Dynamic Range: 93.75-3000 pg/ml
  • Sensitivity: 47 pg/ml
  • Incubation: 1h45
The Eagle Biosciences Human soluble CD95 (sCD95) ELISA is to be used for the in-vitro quantitative determination of soluble CD95 (APO-1, Fas) in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant human sCD95. The Eagle Biosciences Human CD95 ELISA Assay kit is for Research Use Only. 
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty