Would you like to log in again?
Human CD213a2 / IL-13Ra2 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
CA231-K01
102994-080EA 830.38 USD InStock
102994-080
Human CD213a2 / IL-13Ra2 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
The Eagle Bios Human IL-13Rα2 ELISA Assay kit is for Research Use Only.
  • Dynamic Range: 156-5000 pg/ml
  • Sensitivity: 150 pg/ml
  • Incubation: 3h45
The Eagle Biosciences Human IL-13Rα2 ELISA is to be used for the in-vitro quantitative determination of interleukin 13 Receptor (IL-13Rα2, also known as CD213α2) in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant human IL-13Rα2. The Eagle Biosciences Human IL-13Rα2 ELISA Assay kit is for Research Use Only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty