Would you like to log in again?
Calprotectin ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
CAL35-K01
102994-084EA 680 USD InStock
102994-084
Calprotectin ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Calprotectin ELISA detects human calprotectin levels in stool samples and is for research use only.
  • Dynamic Range: 69.5 - 5560 ng/ml
  • Sensitivity: 5 ng/ml
  • Incubation: 2 hours
This Eagle Biosciences Calprotectin ELISA Assay kit is intended for use in the quantitative determination of human calprotectin (neutrophil cytoplasmic protein S100A8/A9, MRP 8/14) levels in stool samples via Enzyme-linked immunosorbent assay. The Calprotectin ELISA Assay kit is for research use only and should not be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty