Would you like to log in again?
Anti-CCP ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
3860
102994-088EA 836.92 USD InStock
102994-088
Anti-CCP ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Anti-CCP EIA detects IgG antibodies against cyclic citrullinated peptides (CCP) in human serum.
  • Dynamic Range: 1 - 2000 U/ml
  • Sensitivity: 1.2 U/ml
  • Incubation: 2 hours
The Eagle Biosciences Anti-CCP ELISA assay kit is designed for determination of IgG autoantibodies to cyclic citrullinated peptides (CCP) in human serum or plasma. Rheumatoid Arthritis (RA) is most often diagnosed by the measurement of rheumatoid factors (RF). Rheumatoid factor is an antibody directed against the Fc-region of IgG. It appears mainly as IgM antibody but also as IgA or IgG subgroup. Rheumatoid factors are present in sera of 70 - 80 % of patients suffering from rheumatoid arthritis. But it is not specific for RA since it will also be found in healthy persons. The incidence depends on the age: for young people it is 1 - 4 %, for older persons it may reach up to 25 %. The advantage of CCP antibodies is a higher sensitivity and specificity for the diagnosis of rheumatoid arthritis in comparison to the rheumatoid factors alone. Anti-CCP is often found at a very early state of the disease and it has a high predictive value for development of the disease. In the first step CCP autoantibodies from the diluted sample (as well as from the calibrators and control) bind to cyclic citrullinated peptides coated on the microtiter plate. After an incubation of 60 minutes at room temperature (RT) unbound components are removed by washing. In a next step bound antibodies reacts with the added anti-human-IgG horseradish peroxidase (HRP) complex. Excessive conjugate is removed after 30 minutes at RT by another washing step. HRP converts the colorless substrate TMB added into a blue product. The enzyme reaction is stopped by adding an acid solution after 15 minutes at RT. The color changes from blue to yellow. The absorbance of the resulting product is measured at 450 / 620 nm within 30 minutes. The obtained OD is direct proportional to the amount of bound antibodies.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty