Would you like to log in again?
Chromogranin A ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
CHR31-K01
102994-098EA 1176.92 USD InStock
102994-098
Chromogranin A ELISA, Eagle Biosciences
Assays ELISAs
Human Chromogranin A ELISA determines human Chromogranin A levels in EDTA-plasma and serum samples.
  • Dynamic Range: 19.2 - 660 ng/ml
  • Sensitivity: 2 ng/ml
  • Incubation: 2.5 hours
The Eagle Biosciences Human Chromogranin A ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of human Chromogranin A levels in EDTA-plasma and serum samples. This assay exclusively measures human Chromogranin A without the high dose “hook” effect up to 1,000,000 ng/ml.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty