Would you like to log in again?
Total Complement Functional Screen ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
COMPL300
102994-102EA 1150.77 USD InStock
102994-102
Total Complement Functional Screen ELISA, Eagle Biosciences
Assays ELISAs
Total Complement Functional Screen ELISA is to determine complement pathways in human serum.
  • Sensitivity: Cut-off
  • Dynamic Range: Cut-off
  • Incubation Time: 2 hours
The complement system plays an essential role in chronic, autoimmune and infectious disease. There are three pathways of complement activation, namely the classical, the alternative and the recently discovered MBL pathway.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty