Would you like to log in again?
Complement Classical pathway ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
COMPLCP310
102994-106EA 1150.77 USD InStock
102994-106
Complement Classical pathway ELISA, Eagle Biosciences, Inc.
Assays ELISAs
The Complement Classical Pathway ELISA is to determine functional classical complement pathway.
  • Sensitivity: Cut-off
  • Dynamic Range: Cut-off
  • Incubation Time: 2 hours
In vitro activation of the complement sequence leads to the consumption of complement components which, in turn, leads to a decrease in their concentration. Thus, the determination of complement proteins or complement activity is used to indicate whether the complement system has been activated by an immunologic and/or pathogenic mechanism. Both functional and immunochemical complement measurements are used to evaluate subjects when a complement-activating disease is suspected or an inherited deficiency is possible. The level of complement activity evaluated by functional assays such as Eagle Biosciences Complement Classical Pathway ELISA Assay Kit  takes into account the rate of synthesis, degradation, and consumption of the components and provides a measure of the integrity of the pathways as opposed to immunochemical methods which specifically measure the concentration of various complement components. 
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty