Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

C-Peptide ELISA, Eagle Biosciences, Inc.

Supplier: Eagle Biosciences
CPT31-K01
102994-116EA 621.15 USD InStock
102994-116
C-Peptide ELISA, Eagle Biosciences, Inc.
Assays ELISAs
C-Peptide ELISA detects human connecting peptide in serum and/or plasma.

  • Dynamic Range: 1.0 - 10 mg/dL
  • Sensitivity: 1.0 mg/dl
  • Incubation: 15 min

C-peptide is a small 31-amino acid peptide usually produced in the beta cell of pancreas as a byproduct of the cleavage of proinsulin in the synthesis of insulin. Proinsulin consists of A and B chain and connecting peptide in the middle, called C-peptide. It is generally found in equimolar amounts equal to insulin in circulation. Since the half-life of C-peptide is 3-4 times that of insulin, it serves as a useful measure of insulin production in the beta cells of the pancreas.

Testing for C-peptide levels can help find the cause of low blood sugar (hypoglycemia) aid in distinguishing type 1 from type 2 diabetes. A person with diabetes may have a normal level of C-peptide which indicates the body is making plenty of insulin but the body is just not responding properly to it. This is the hallmark of type 2 diabetes (adult insulin-resistant diabetes). For subjects with type 1 diabetes treated with insulin, measuring C-peptide level is useful in evaluating beta cell function related to synthesis and release endogenous insulin into the circulation. Some studies have suggested that C-peptide may have chemotactic effects on the inflammatory cells and might have a role in increased risk of atherosclerosis in persons with type-2 diabetes.

Assay Principle

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty