Would you like to log in again?
C-Peptide Ultrasensitive ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
CPU31-K01
102994-118EA 836.92 USD InStock
102994-118
C-Peptide Ultrasensitive ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Human C-peptide Ultra ELISA is intended to detect human c-peptide in serum or plasma samples.
  • Dynamic Range: 8.6 pg/ml- 700 pg/ml
  • Sensitivity: 0.57 pg/ml
  • Incubation: 2.5 hours
C-peptide is a small 31-amino acid peptide usually produced in the beta cell of pancreas as a byproduct of the cleavage of proinsulin in the synthesis of insulin. Proinsulin consists of A and B chain and connecting peptide in the middle, called C-peptide. It is generally found in equimolar amounts equal to insulin in circulation. Since the half-life of C-peptide is 3-4 times that of insulin, it serves as a useful measure of insulin production in the beta cells of the pancreas.
Testing for C-peptide levels can help find the cause of low blood sugar (hypoglycemia) aid in distinguishing type 1 from type 2 diabetes. A person with diabetes may have a normal level of C-peptide which indicates the body is making plenty of insulin but the body is just not responding properly to it. This is the hallmark of type 2 diabetes (adult insulin-resistant diabetes). For subjects with type 1 diabetes treated with insulin, measuring C-peptide level is useful in evaluating beta cell function related to synthesis and release endogenous insulin into the circulation.
Some studies have suggested that C-peptide may have chemotactic effects on the inflammatory cells and might have a role in increased risk of atherosclerosis in persons with type-2 diabetes.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty