Would you like to log in again?
Creatinine Assay Kit (urine normalizing assay), Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
CRE34-K01
102994-120EA 255 USD InStock
102994-120
Creatinine Assay Kit (urine normalizing assay), Eagle Biosciences, Inc.
Assays ELISAs
Creatinine Microplate is to detect Creatinine in urine by a microplate .
  • Dynamic Range: 5 - 80 AU/ml
  • Sensitivity: 0.09 AU/ml
  • Incubation: 2.5 hours
Creatine (Cr) is produced in the kidney, liver and pancreas, phosphorylated, and transported to the brain and muscle tissue. However, a small proportion of free Cr is converted irreversibly to creatinine (Crn) in the muscular tissue in proportion to the muscle mass. The amount of Crn excreted daily by an individual is relatively constant. Thus, urinary creatinine levels may be used as an index of standardization. 24-h urinary Crn excretion is used to estimate total muscle mass because the rate of non-enzymatic production of Crn from Cr is nearly constant and >90% of the total body Cr is found in muscle tissue. Normal urinary creatinine values for men and women range from 9.7 – 24.7 and 7.9 – 14.2 mmol/24h respectively. Changes in excretion rate may be indicative of impaired renal metabolism.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty