Would you like to log in again?
C-Reactive Protein (CRP) ELISA , Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
CRP31-K01
102994-122EA 568.85 USD InStock
102994-122
C-Reactive Protein (CRP) ELISA , Eagle Biosciences, Inc.
Assays ELISAs
hsCRP ELISA is intended to detect C-reactive protein (CRP) in human serum samples.
  • Dynamic Range: 0.62mg/l - 119.3 mg/l
  • Sensitivity: 0.1 mg/l
  • Incubation: 1.5 hours
C-Reactive Protein (CRP) has been demonstrated to be a general indicator of major tissue damage. Hence, studies have shown that CRP can be used to indicate a stroke or heart attack because major blood vessels leading to the heart or brain are damaged and release large quantities of CRP during these disease states. CRP is a particularly useful indicator of CHD in women and in patients that demonstrate no other plasma circulating biochemical indicators.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty