Would you like to log in again?
Hydroxypyridinium Crosslinks HPLC Assay (100 Tests), Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
CSL34-H100
102994-128EA 1235.77 USD InStock
102994-128
Hydroxypyridinium Crosslinks HPLC Assay (100 Tests), Eagle Biosciences, Inc.
Assays Immunological Assays
The Hydroxypyridinium Crosslinks HPLC Assay is used to detect hydroxypyridinium crosslinks in urine
  • Incubation Time: 4 hours 20 minutes
  • Sample Size: 1 mL
  • Sample Type: Urine
Pyridinium and deoxypyridinium are built as intramolecular crosslinkers during the maturation of collagen. During bone resorption collagen is degraded and the pyridinium-crosslinks are excreted. They are not reutilized by the body and excreted in the urine. The crosslinks are not released during the de novo synthesis of collagen. They are specific for resorptive processes. Neither PYD nor DPD are taken up by nutrition. Therefore the determination of the crosslinks is independent from the diet. PYD is found in cartilage, ligaments and bone, whereas DPD is rather specific for bone. The crosslink excretion shows a strong circadian rhythm with a maximum in the early morning hours. Reliable data can be obtained with a 24h or a creatinine corrected morning urine. An elevated crosslink excretion is shown in osteoporotic women. Crosslink levels decrease during treatment with estrogen (Seibel et al. 1994). Therefore they are suited for therapy monitoring (Ganero et al. 1994). The measurement of pyridinium crosslinks is an independent predictor for future bone fractures (Ganero et al. 1995). Increased crosslink concentrations are also described in patients with Paget’s Disease, primary hyperparathyroidism and bone metastasis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty