Would you like to log in again?
Anti-CaSR IgG ELISA _ DO NEW PROTOCOL, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
CSR31-K01
102994-130EA 1268.46 USD InStock
102994-130
Anti-CaSR IgG ELISA _ DO NEW PROTOCOL, Eagle Biosciences, Inc.
Assays ELISAs
Human Anti-CaSR IgG EIA detects human anti-CaSR IgG autoAb levels in serum, plasma, tissue extract.
  • Dynamic Range: 10 - 270 U/ml
  • Sensitivity: 2.5 U/ml
  • Incubation: 2.0 hours
The Eagle Biosciences Human Anti-CaSR ELISA Assay Kit (enzyme-linked immunoassay kit) kit is produced for the quantitative determination of human anti-CaSR (calcium sensing receptor) autoantibody levels in serum, plasma, tissue extract or other liquid samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty