Would you like to log in again?
CXCL 12/SDF-1 Alpha ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
CXC31-K01
102994-140EA 1340.38 USD InStock
102994-140
CXCL 12/SDF-1 Alpha ELISA, Eagle Biosciences
Assays ELISAs
CXCL 12/SDF-1 Alpha ELISA is used to determine CXCL 12 / SDF-1 Alpha in serum and plasma.
  • Dynamic Range: 0.625 - 40 ng/ml
  • Sensitivity: 0.31 ng/ml
  • Incubation: 2.5 hours
SDF-1 Alpha (CXCL12) is a CXC chemokine [1]. The small basic protein (ca. 8 kDa) has no glycosylation site in its sequence [2]. The homology properties of SDF-1 Alpha are unique among the chemokines. It has a high degree of conservation within the mammalians, with 99 % identity between the human and the murine SDF-1 Alpha sequences. But it shows a weak homology to the other CXC chemokines and the SDF-1 gene is on chromosome 10, whereas the other chemokine genes are on chromosome 4 and 17 [3].
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty