Would you like to log in again?
Anti-dsDNA ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
4015
102994-260EA 568.85 USD InStock
102994-260
Anti-dsDNA ELISA, Eagle Biosciences
Assays ELISAs
Anti-dsDNA ELISA determines IgG antibodies to double-stranded desoxyribonucleic acid in serum.
  • Dynamic Range: 1-300 IU/mL
  • Sensitivity: 1 IU/mL
  • Incubation: 2.5 hours
The Eagle Biosciences Anti-dsDNA ELISA Assay Kit is used for the quantitative determination of IgG antibodies to double-stranded desoxyribonucleic acid (dsDNA) in human serum for the diagnosis of systemic lupus erythematosus (SLE). The kit is for research use only and not for diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty