Would you like to log in again?
hGH Sensitive ELISA , Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
E022
102994-264EA 882.69 USD InStock
102994-264
hGH Sensitive ELISA , Eagle Biosciences, Inc.
Assays ELISAs
hGH Sensitive ELISA is quantitatively measures levels of human growth hormone in serum and plasma.
  • Dynamic Range: 0.05 - 1.0 ng/mL
  • Sensitivity: 12 pg/ml
  • Incubation: 2.5 hours
The Endocrinology system of human Growth Hormone (hGH), also named Somatropin, is characterized by an extreme complexity. hGH is the product of the GH-1 gene located on chromosome 17 and expressed in pituitary cells. 80% of the gene expression results in a non-glycolsylated 22 kDa protein consisting of 191 amino acids. The other 20% of gene expression results in a variant form of 20 kDa by alternative splicing. Additionally, several smaller variants can be found in circulation as well as translational modified proteins and different degrees of protein aggregation. Further on, bioactivity of Growth Hormone is regulated by a specific binding protein (GHBP) formed by the extra cellular part of the cellular transmembran GH-receptor. These modifications allow a tight control of the half-life period hGH and of its bioactivity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty