Would you like to log in again?
Mouse-/Rat-GH ELISA , Eagle Biosciences
Supplier: Eagle Biosciences
E023
102994-266EA 921.92 USD InStock
102994-266
Mouse-/Rat-GH ELISA , Eagle Biosciences
Assays ELISAs
Mouse/Rat GH ELISA is used to detect concentration of Growth Hormone in serum and plasma samples.
  • Dynamic Range: 0.025 – 1.5 ng/mL
  • Sensitivity: 0.025 ng/ml
  • Incubation: 3 hours
The Eagle Biosciences Mouse/Rat GH ELISA Assay Kit is intended to be used for the measurement of Growth Hormone in mouse and rat serum and plasma samples. The Eagle Biosciences Mouse/Rat GH ELISA Assay Kit is for Research Use Only. 
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty