Would you like to log in again?
GHBP ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
E024
102994-268EA 921.92 USD InStock
102994-268
GHBP ELISA, Eagle Biosciences, Inc.
Assays ELISAs
GHBP ELISA is used to measure human Growth Hormone Binding Protein levels in serum or plasma.
  • Dynamic Range: 0.05 – 4ng/mL,
  • Sensitivity: 0.008 ng/mL
  • Incubation: 3 hours
Hormone Binding Protein (GHBP) consists of 238 amino acids and includes four sides for glycosylation and three disulfide bounds. In humans GHBP is formed by receptor shedding of the growth hormone receptor by a metalloprotease (ADAM17).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty