Would you like to log in again?
functional IGFBP-3 LIA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
E04A
102994-274EA 882.69 USD InStock
102994-274
functional IGFBP-3 LIA, Eagle Biosciences, Inc.
Assays Immunological Assays
The Eagle Biosciences IGFBP-3 LIA utilizes specific and high affinity antibodies for IGFBP-3.
  • Sensitivity: 0.4 ng/mL
  • Dynamic Range: 0.4 - 30 ng/mL
  • Incubation Time: 3.5 hours
All currently existing IGFBP-3 immunoassays use the binding of specific anti-IGFBP-3 antibodies for signal generation and thus IGFBP-3 quantification. The failure of differentiation between complete IGFBP-3 molecules and their respective fragments (derived physiologically due to the different proteases activities) is unavoidable in this system. Because one molecule IGFBP-3 can be cleaved in several fragments often false high quantitative values are measured. Based on this methodology it is not possible to differentiate between high IGFBP-3 levels in fact, or, a high degree of fragmentation. The incidental attempts to use monoclonal antibodies with a binding region represented only by the intact IGFBP-3 molecule are indirect, imprecise and insufficient. The activities of all effective proteases, which have different sites of action and therefore generate different kind of fragments, are disregarded.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty