Would you like to log in again?
IGFBP-2 ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
E05
102994-276EA 921.92 USD InStock
102994-276
IGFBP-2 ELISA, Eagle Biosciences
Assays ELISAs
IGFBP-2 ELISA detects human IGFBP-2 in biological samples
  • Dynamic Range: 2 -80 ng/ml
  • Sensitivity: 0.2 ng/ml
  • Incubation: 2 hours
Insulin-like growth factors (IGFs) regulate the proliferation, differentation, apoptose, cell adhesion and metabolism in various tissues and cell types. The IGF-I, which is produced mainly in liver under the influence of growth hormone (GH), regulates as hormone the linear growth of the bones and the process of sexual maturity, while IGF-II is mainly a growth factor of fetal tissue. The biological actions of IGF over the IGF-Type-I receptor are modulated variably through the IGF binding proteins (IGFBP-1 to-6). IGFBP-2 is, after IGFBP-3, the second most frequent IGFBP in the human blood. IGFs, especially tumor typical pro-IGF-forms and hormones regulate the expression of IGFBP-2, GH effect is thereby inhibiting. At cellular level IGFBP-2 seems to stimulate the proliferation and dissemination of solid tumors via an IGF-independent mechanism.
For Research Use Only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty