Would you like to log in again?
Mouse-/ Rat-LEPTIN ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
E06
102994-278EA 830.38 USD InStock
102994-278
Mouse-/ Rat-LEPTIN ELISA, Eagle Biosciences
Assays ELISAs
Leptin Sensitive ELISA measures human Leptin in various samples.
  • Dynamic Range: 0.75-24 U/ml
  • Sensitivity: 0.2 U/ml
  • Incubation: 1h40
Leptin, the product of the ob gene (1,2), is a recently discovered single-chain proteohormone with a molecular weight of 16 kD which is thought to play a key role in the regulation of body weight. Its amino acid sequence exhibits no major homologies with other proteins (1). Leptin is almost exclusively produced by differentiated adipocytes (3-5). It acts on the central nervous system, in particular the hypothalamus, thereby suppressing food intake and stimulating energy expenditure (2,6-9). Leptin receptors - alternatively spliced forms exist that differ in length - belong to the cytokine class I receptor family (10-12). They are found ubiquitously in the body (10,11,13,14) indicating a general role of leptin which is currently not fully understood. A circulating form of the leptin receptor exists which acts as one of several leptin binding proteins (15).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty