Would you like to log in again?
Mouse- / Rat-IGFBP-2 ELISA , Eagle Biosciences
Supplier: Eagle Biosciences
E08
102994-282EA 921.92 USD InStock
102994-282
Mouse- / Rat-IGFBP-2 ELISA , Eagle Biosciences
Assays ELISAs
Mouse/Rat IGFBP-2 ELISA is suited for the measuring of IGFBP-2 in the mouse and the rat serum.
  • Dynamic Range: 0.01 - 200 ng/mL
  • Sensitivity: 0.01 ng/mL
  • Incubation: 3 hours
Insulin-like growth factors (IGFs) regulate the proliferation, differentation, apoptosis, cell adhesion and metabolism in various tissues and cell types IGFBP-2 is an unglycosylated polypetide of 31.3 kDa, which forms binary IGF-complexes and shows no circadian rhythm in the circulation. The serum concentration of IGFBP-2 increases in fasting, after major surgery and after trauma, but the increasing of the concentration is most intensive in malignant diseases. The correlation of the IGFBP-2 level to the degree of progression is a striking feature in various tumor types as is the normalization of the IGFBP-serum levels after remission (5-10). During the GH-therapy, e.g. in short stature and in GH-abuse (doping) the IGFBP-2 level decreases. In Trisomy 18 IGFBP-2 in maternal serum is decreased and IGFBP-1 is increased; therefore the ratio IGFBP-2 /IGFBP-1 is a marker for this chromosome abnormality (17). Transgenic organisms are a good opportunity to investigate the function of genes or proteins. The mouse or rat model is a well-suited system for investigation of the relevance of IGFBP-2 in physiological and pathological processes. Over expression of the IGFBP-2 gene in mice results in a weight reduction of 30% in spleen and moderately reduced weight in other organs (18). Effects of IGFBP-2 on the organism can be compensated through the modified expression of other IGF Binding proteins. Especially in tumor biology the mouse and rat systems enable investigation of the systemic relevance of IGFBP-2. IGFBP-2 influences tumor cells as it induces catalase activity in adrenocortical cells (19). Furthermore IGFBP-2 interacts with tumor cells via its RGD-amino acid sequence and seems to stimulate cell invasion of glioma cells (20).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty