Would you like to log in again?
ADIPONECTIN ELISA , Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
E09
102994-284EA 738.85 USD InStock
102994-284
ADIPONECTIN ELISA , Eagle Biosciences, Inc.
Assays ELISAs
Adiponectin ELISA measures Adiponectin in the human serum & plasma samples for research use only.
  • Dynamic Range: 2 -100 ng/ml
  • Sensitivity: 0.6 ng/ml
  • Incubation: 1.75 hours
Adiponectin is a 30kDa protein which percentage in serum proteins is 0.01%. It is mainly synthesized by Adipocytes, but also muscle cells and hepatocytes have the ability to synthesize Adiponectin. Until now, IGF-I is the only known natural inductor of the synthesis. It consists of  a Collagen-like N-terminal and a globular C-terminal domain [1]. In vivo Adiponectin appears with different oligomers. Beside the trimer and ditrimer also high molecular multimers exist [1-3]. Up to now two different receptors are known, both receptors are ubiquitary expressed, though the distribution in the tissues varies. The Adiponectin Receptor 1 (AdipoR1) is especially in muscle- and AdipoR2 in liver tissue synthesized [4].
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty