Would you like to log in again?
Mouse-ADIPONECTIN ELISA , Eagle Biosciences
Supplier: Eagle Biosciences
E091-M
102994-286EA 797.69 USD InStock
102994-286
Mouse-ADIPONECTIN ELISA , Eagle Biosciences
Assays ELISAs
Mouse Adiponectin ELISA measures the Adiponectin in the Mouse serum and the plasma samples.
  • Dynamic Range: 0.025 - 1 ng/mL
  • Sensitivity: 0.008 ng/mL
  • Incubation: 2.5 hours
Adiponectin was described for the first time in the early 90th of the last century as an Endocrinology factor produced by adipocytes. Adiponectin is involved in regulation of energy- and fat metabolism. So its concentration in the circulation is said to reflect the risk of atherosclerosis and the degree of insulin resistance. Based on the high incidence of these diseases, adiponectin was and still is object of intensive research regarding the underlying biological mechanisms and regarding its value as biomarker. Beside different cell culture models and studies with human patients, mice and rats are suitable model organisms for basic research and pre-clinical studies.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty