Would you like to log in again?
Anti-Myelin Auto-Antibodies ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
E100
102994-292EA 2798.46 USD InStock
102994-292
Anti-Myelin Auto-Antibodies ELISA, Eagle Biosciences
Assays ELISAs
Anti-Myelin Auto-Antibodie EIA detects antigens IgG and IgM Ab in serum samples&cerebrospinal fluid.
  • Sample Size: 10uL
  • Sensitivity: 0.014 ng/ml
  • Incubation: 4.75 hours
Several neurodegenerative diseases are at least partially caused by auto immunreactions. Inflammatory processes result in neuronal loss. Mostly, T-cell dependent immunity is in the central part of the pathological concept. But it is known that in some neurological diseases like multiple sclerosis B-cell dependent immunity is also of relevance in disease progress.  Thus, antibodies are a well established means in differential diagnostics of neuronal diseases e.g. by the detection of oligoclonal bands or measurement of specific antibodies against neuronal Proteins like aquaporin IV.   This assay allows the quantitative detection of Antigens specific IgG and IgM antibodies in serum samples and cerebrospinal fluid.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty