Would you like to log in again?
PROGRANULIN ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
E103
102994-296EA 882.69 USD InStock
102994-296
PROGRANULIN ELISA, Eagle Biosciences
Assays ELISAs
Progranulin ELISA is to be used for the determination of Progranulin in serum or plasma samples.
  • Dynamic Range: 75-2500 pg/mL
  • Sensitivity: 18 pg/mL
  • Incubation: 2 hours
Progranulin is also known as Granulin Epithelin Precursor, Proepithelin or Acrogranin. It is a 68.5 kDa protein, consisting of 593 amino acids (inclusive Signalpeptid), which appears in vivo in strongly glycosylated form and therefore has a size of approximately 90 kDa (1).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty