Would you like to log in again?
Anti-HAV IgM ELISA , Eagle Biosciences
Supplier: Eagle Biosciences
E11
102994-300EA 830.38 USD InStock
102994-300
Anti-HAV IgM ELISA , Eagle Biosciences
Assays ELISAs
Anti-HAV IgM ELISA detects IgM Ab in human serum or plasma against the Hepatitis A-Virus.
  • Dynamic Range: Cutoff
  • Sensitivity: Cutoff
  • Incubation: 3.5 hours
Positive detection of antibodies directed against the Hepatitis A virus (anti-HAV) is evidence of immunity to Hepatitis-A virus (1). After natural infection with the Hepatitis A-Virus, neutralising antibodies appear at the same time of Anti-HAV of IgG-Class formation (1).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty