Would you like to log in again?
Anti-Pseudomonas aeruginosa ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
E15
102994-304EA 3040.38 USD InStock
102994-304
Anti-Pseudomonas aeruginosa ELISA, Eagle Biosciences
Assays ELISAs
Anti-Pseudomonas aeruginosa IgG ELISA detects IgG-Ab in human serum against extracellular proteins.
  • Dynamic Range: Cutoff
  • Sensitivity: 0.02 ng/ml
  • Incubation: 4.75 hours
Pseudomonas aeruginosa, a Gram-negative bacterium ubiquitously distributed in the moist environment, causes about 10% of all nosocomial infections. This opportunistic pathogen leads to acute and chronic types of infection within the various organs.  P. aeruginosa infection provokes a rapid production of antibodies to a large number of P. aeruginosa Antigenss. Eagle Biosciences’s sensitive antibody detection system detects a P.aeruginosa infection very early at the onset.  Due to the use of three P.aeruginosa Antigenss, which are highly immunogenic and present in different parts from nearly all P.aeruginosa strains.  Depending on the Pseudomonas aeruginosa species and the immune reaction, antibodies can be detected against a single, two or even all three Antigenss simultaneously. A sample is regarded as sero-positive when antibodies against one or more of the Antigenss can be detected.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty